Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminopropyltriethoxysilane, 99%

Catalog No. AC151080025
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC151080025 2.5 kg Glass bottle
AC151080050 5 g Glass bottle
AC151081000 100 g Glass bottle
AC151085000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC151080025 Supplier Thermo Scientific Chemicals Supplier No. 151080025
Only null left
Add to Cart

CAS: 919-30-2 | C9H23NO3Si | 221.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 919-30-2
Molecular Formula C9H23NO3Si
Molecular Weight (g/mol) 221.37
MDL Number MFCD00008207,MFCD01324904
InChI Key WYTZZXDRDKSJID-UHFFFAOYSA-N
Synonym 3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane
PubChem CID 13521
IUPAC Name 3-triethoxysilylpropan-1-amine
SMILES CCO[Si](CCCN)(OCC)OCC

Specifications

Melting Point -70.0°C
Density 0.9420g/mL
Boiling Point 217.0°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4200 to 1.4220
Linear Formula H2N(CH2)3Si(OC2H5)3
Quantity 2.5 kg
Beilstein 04,IV,4273
Specific Gravity 0.942
Solubility Information Solubility in water: reacts. Other solubilities: soluble in chloroform
Formula Weight 221.37
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Aminopropyltriethoxysilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 213-048-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.