Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminopropyltriethoxysilane, 99%, ACROS Organics™

missing translation for 'orderingAttributeHoverText'
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
919-30-2
C9H23NO3Si
221.37
WYTZZXDRDKSJID-UHFFFAOYSA-N
3-aminopropyltriethoxysilane,3-aminopropyl triethoxysilane,aptes,3-triethoxysilyl propan-1-amine,1-propanamine, 3-triethoxysilyl,silicone a-1100,silane 1100,3-triethoxysilyl propylamine,propylamine, 3-triethoxysilyl,triethoxy 3-aminopropyl silane
13521
3-triethoxysilylpropan-1-amine
CCO[Si](CCCN)(OCC)OCC
919-30-2
C9H23NO3Si
221.37
WYTZZXDRDKSJID-UHFFFAOYSA-N
3-aminopropyltriethoxysilane,3-aminopropyl triethoxysilane,aptes,3-triethoxysilyl propan-1-amine,1-propanamine, 3-triethoxysilyl,silicone a-1100,silane 1100,3-triethoxysilyl propylamine,propylamine, 3-triethoxysilyl,triethoxy 3-aminopropyl silane
13521
3-triethoxysilylpropan-1-amine
CCO[Si](CCCN)(OCC)OCC
Specific Gravity 0.942
Density 0.9420g/mL
Refractive Index 1.4200 to 1.4220
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point -70.0°C
Quantity 100g
Boiling Point 217.0°C
Color Undesignated
Flash Point 96°C
Assay Percent Range 99%
Linear Formula H2N(CH2)3Si(OC2H5)3
Beilstein 04,IV,4273
Solubility Information Solubility in water: reacts. Other solubilities: soluble in chloroform
Formula Weight 221.37
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 3-Aminopropyltriethoxysilane
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.