Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminopyrazole-4-carboxamide Hemisulfate 98.0+%, TCI America™
SDP

Catalog No. A153225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A153225G 25 g
A15325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A153225G Supplier TCI America Supplier No. A153225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27511-79-1
Molecular Formula C8H14N8O6S
Molecular Weight (g/mol) 350.31
MDL Number MFCD00013093
InChI Key UMPKASYMNORSRO-UHFFFAOYSA-N
Synonym 3-amino-4-pyrazolecarboxamide hemisulfate, 5-aminopyrazole-4-carboxamide hemisulphate, 3-amino-1h-pyrazole-4-carboxamide hemisulfate, 1h-pyrazole-4-carboxamide, 3-amino-, sulfate 2:1, 3-amino-4-pyrazolecarboxamide hemisulfate salt, 3-aminopyrazole-4-carboxamide hemisulfate, 5-amino-1h-pyrazole-4-carboxamide; sulfuric acid, bis 3-amino-1h-pyrazole-4-carboxamide ; sulfuric acid, bis 3-amino-2h-pyrazole-4-carboxamide ; sulfuric acid, 5-amino-1h-pyrazole-4-carboxamide hemisulphate
PubChem CID 2723907
IUPAC Name 5-amino-1H-pyrazole-4-carboxamide;sulfuric acid
SMILES C1=NNC(=C1C(=O)N)N.C1=NNC(=C1C(=O)N)N.OS(=O)(=O)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 175.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminopyrazole-4-carboxamide Hemisulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.