Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminopyridine 99.0+%, TCI America™
SDP

Catalog No. A0413500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0413500G 500 g
A041325G 25 g
A0413100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0413500G Supplier TCI America Supplier No. A0413500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 462-08-8 | C5H6N2 | 94.12 g/mol

Chemical Identifiers

CAS 462-08-8
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.12
MDL Number MFCD00006400
InChI Key CUYKNJBYIJFRCU-UHFFFAOYSA-N
Synonym 3-aminopyridine, 3-pyridinamine, 3-pyridylamine, pyridin-3-ylamine, m-aminopyridine, amino-3 pyridine, pyridine, 3-amino, 3-amino pyridine, beta-aminopyridine, amino-3-pyridine
PubChem CID 10009
IUPAC Name pyridin-3-amine
SMILES NC1=CC=CN=C1

Specifications

Melting Point 62°C
pH 10 (100g/L H2O soln. 20°C)
Boiling Point 248°C
Quantity 500 g
UN Number 2671
Formula Weight 94.12
Percent Purity ≥99.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminopyridine
EINECSNumber (5)-0725
RTECSNumber US1650000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.