Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminoquinuclidine Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. A123625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A123625G 25 g
A12365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A123625G Supplier TCI America Supplier No. A123625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6530-09-2
Molecular Formula C7H16Cl2N2
Molecular Weight (g/mol) 199.12
MDL Number MFCD00137395
InChI Key STZHBULOYDCZET-UHFFFAOYNA-N
Synonym 3-aminoquinuclidine dihydrochloride, 1-azabicyclo 2.2.2 octan-3-amine dihydrochloride, quinuclidine, 3-amino-, dihydrochloride, 3aminoquinuclidine dihydrochloride, quinuclidin-3-amine dihydrochloride, 1-aza-bicyclo 2.2.2 oct-3-ylamine dihydrochloride, 1-azabicyclo 2.2.2 octan-3-amine, dihydrochloride, 3-amino-1-azabicyclo 2.2.2 octane dihydrochloride, quinuclidine-3-ylamine, chloride, chloride
PubChem CID 197853
IUPAC Name 1-azabicyclo[2.2.2]octan-3-amine;dihydrochloride
SMILES C1CN2CCC1C(C2)N.Cl.Cl

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 199.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminoquinuclidine Dihydrochloride
RTECSNumber VD4590000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.