Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Anilino-1-propanol 98.0+%, TCI America™
SDP

Catalog No. A224825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A224825G 25 g
A22485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A224825G Supplier TCI America Supplier No. A224825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 31121-11-6
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD15206074
InChI Key GZYBSURBYCLNSM-UHFFFAOYSA-N
Synonym N-(3-Hydroxypropyl)aniline
PubChem CID 410467
IUPAC Name 3-(phenylamino)propan-1-ol
SMILES OCCCNC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 140°C
Quantity 25 g
Formula Weight 151.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Anilino-1-propanol
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.