Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Azido-3'-deoxythymidine, 98%

Catalog No. AC454980050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454980050 5 g
AC454980010 1 g
AC454980250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC454980050 Supplier Thermo Scientific Chemicals Supplier No. 454980050
Add to Cart

CAS: 30516-87-1 | C10H13N5O4 | 267.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30516-87-1
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.25
MDL Number MFCD00006536
InChI Key HBOMLICNUCNMMY-CFQLRCIDNA-N
Synonym azt; azidothymidine; zdv; zidovudine, 1-2r-4-azido-5-hydroxymethyl oxolan-2-yl-5-methyl-3h-pyrimidine-2,4-dione
PubChem CID 455007
IUPAC Name 1-[(2R,5S)-4-azido-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](CO)O2)C(=O)NC1=O

Specifications

Melting Point 118.0°C to 120.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Merck Index 15,10322
Formula Weight 267.24
Percent Purity 98%
Chemical Name or Material 3'-Azido-3'-deoxythymidine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Suspected of causing cancer. May cause cancer by inhalation. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.