Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Benzyloxyacetophenone 98.0+%, TCI America™
SDP

Catalog No. B470525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B470525G 25 g
B47055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B470525G Supplier TCI America Supplier No. B470525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34068-01-4
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.275
MDL Number MFCD00026221
InChI Key FGQMEAWGAUALJQ-UHFFFAOYSA-N
Synonym 1-3-benzyloxy phenyl ethanone, 3-benzyloxyacetophenone, 3'-benzyloxyacetophenone, 1-3-benzyloxy phenyl ethan-1-one, 1-3-phenylmethoxy phenyl ethan-1-one, 3'-benzyloxy acetophenone, ethanone, 1-3-phenylmethoxy phenyl, 1-acetyl-3-phenylmethoxy benzene
PubChem CID 98689
IUPAC Name 1-(3-phenylmethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2

Specifications

Color White-Yellow
Boiling Point 154°C
Quantity 25 g
Formula Weight 226.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3′-Benzyloxyacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.