Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Benzyloxybenzyl Alcohol 97.0+%, TCI America™
SDP

Catalog No. B385725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B385725G 25 g
B38575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B385725G Supplier TCI America Supplier No. B385725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1700-30-7
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.264
MDL Number MFCD00004642
InChI Key AFKLSWIRJUJWKY-UHFFFAOYSA-N
Synonym 3-benzyloxybenzyl alcohol, 3-benzyloxy phenyl methanol, 3-benzyloxybenzylalcohol, benzenemethanol, 3-phenylmethoxy, 3-phenylmethoxyphenyl methanol, 3-benzyloxyphenyl methanol, 3-phenylmethoxy phenyl methan-1-ol, pubchem7731, acmc-1bo1a
PubChem CID 74341
IUPAC Name (3-phenylmethoxyphenyl)methanol
SMILES C1=CC=C(C=C1)COC2=CC=CC(=C2)CO

Specifications

Melting Point 49°C
Boiling Point 160°C
Quantity 25 g
Formula Weight 214.26
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Benzyloxybenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.