Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Boc-amino)azetidine, 98%

Catalog No. aah59556me
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59556ME 500 mg
AAH59556MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59556ME Supplier Thermo Scientific Chemicals Supplier No. H59556ME
Only null left
Add to Cart

CAS: 91188-13-5 | C8H16N2O2 | 172.23 g/mol

It is an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate in organic syntheses.

Solubility
Soluble in water. (5g/100ml)

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91188-13-5
Molecular Formula C8H16N2O2
Molecular Weight (g/mol) 172.23
MDL Number MFCD01861759
InChI Key NEMXVXVJGXZDRR-UHFFFAOYSA-N
Synonym tert-butyl azetidin-3-ylcarbamate, 3-n-boc-amino-azetidine, 3-boc-aminoazetidine, 3-n-boc-amino azetidine, tert-butyl n-azetidin-3-yl carbamate, 3-n-boc-aminoazetidine, 3-boc-amino azetidine, azetidin-3-yl-carbamic acid tert-butyl ester, 3-boc-amino-azetidine, 3-tert-butoxycarbonylamino azetidine
PubChem CID 1519405
IUPAC Name tert-butyl N-(azetidin-3-yl)carbamate
SMILES CC(C)(C)OC(=O)NC1CNC1

Specifications

Boiling Point 268°C
Solubility Information Soluble in water. (5g/100ml)
Density 1.06
Quantity 500 mg
Flash Point 116°C (241°F)
Formula Weight 172.23
Percent Purity 98%
Chemical Name or Material 3-(Boc-amino)azetidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.