Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Boc-aminomethyl)azetidine hydrochloride, 95%

Catalog No. AAH59816MD
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59816MD 250 mg
AAH59816ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59816MD Supplier Thermo Scientific Chemicals Supplier No. H59816MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 91188-15-7 | C9H18N2O2 | 186.26 g/mol

It is a useful building block in the synthesis of polypeptides and other nitrogen containing compounds with potential biological properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful building block in the synthesis of polypeptides and other nitrogen containing compounds with potential biological properties.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91188-15-7
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.26
MDL Number MFCD01861760
InChI Key MOLUHRBHXXGWDP-UHFFFAOYSA-N
Synonym 3-n-boc-aminomethyl azetidine, 3-boc-aminomethylazetidine, tert-butyl n-azetidin-3-ylmethyl carbamate, tert-butyl azetidin-3-ylmethyl carbamate, azetidin-3-ylmethyl-carbamic acid tert-butyl ester, tert-butyl azetidin-3-ylmethylcarbamate, 3-boc-aminomethyl-azetidine, 3-n-boc aminomethylazetidine, tert-butyl azetidin-3-yl methyl carbamate, 3-boc-3-aminomethylazetidine
PubChem CID 15346872
SMILES CC(C)(C)OC(=O)NCC1CNC1

Specifications

Solubility Information Slightly soluble in water.
IUPAC Name tert-butyl N-(azetidin-3-ylmethyl)carbamate
Quantity 250 mg
Formula Weight 222.71
Percent Purity 95%
Chemical Name or Material 3-(Boc-aminomethyl)azetidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.