Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-3-(Boc-aminomethyl)piperidine, 98%, Thermo Scientific™

Catalog No. AAH6025703
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6025703 1 g
AAH60257MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6025703 Supplier Thermo Scientific Chemicals Supplier No. H6025703

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142643-29-6
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD01632454
InChI Key KHPQHXGYYXYTDN-UHFFFAOYSA-N
Synonym 3-n-boc-aminomethylpiperidine, tert-butyl piperidin-3-ylmethyl carbamate, 3-boc-aminomethyl-piperidine, 3-n-boc-aminomethyl piperidine, 3-boc-aminomethylpiperidine, tert-butyl n-piperidin-3-ylmethyl carbamate, piperidin-3-ylmethyl-carbamic acid tert-butyl ester, tert-butyl n-3-piperidylmethyl carbamate, tert-butyl piperidin-3-ylmethylcarbamate, tert-butyl piperidin-3-yl methylcarbamate
PubChem CID 2756030
IUPAC Name tert-butyl N-(piperidin-3-ylmethyl)carbamate
SMILES CC(C)(C)OC(=O)NCC1CCCNC1

Specifications

Melting Point 60°C to 70°C
UN Number UN3259
Beilstein 7023643
Quantity 1 g
Formula Weight 214.3
Percent Purity 98%
Chemical Name or Material (+/-)-3-(Boc-aminomethyl)piperidine
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
Recommended Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.