Learn More
Sartorius 3 Branch Microsart™ Manifolds
Catalog No. 14558829 Shop All Sartorius Products
Change view
Click to view available options
Type:
Biosart 100 Manifold
Biosart 250 Manifold
Microsart Manifold
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-558-829 Biosart 250 Manifold
14-558-826 Biosart 100 Manifold
14-558-822 Microsart Manifold
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-558-829 Supplier Sartorius Supplier No. 168M3BS250
Only null left
Add to Cart
Add to Cart
Filtration stand designed to adapt to a variety of different needs.
  • 100% stainless steel for minimized cleaning effort
  • Full autoclavability without disassembly
  • Sterile venting in critical applications
  • Reduced risk of secondary contamination
  • Quick connection adapters for fast connection of tubing and fast interconnection of 2 manifolds
  • Low working height for ergonomic working in laminar flow
  • Accurately detects and quantifies microorganisms in liquid samples
  • Method of choice for liquid testing
  • Provides reliable and reproducible results
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
Filter Type Manifold
For Use With (Equipment) Biosart 250 Funnel
Material Stainless Steel
Sterility Non-sterile
Quantity 1
Type Biosart 250 Manifold
Volume (Metric) 250 mL
Length (Metric) 474 mm
Width (Metric) 98 mm
Height (Metric) 120 mm