Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 3-BrB-PP1, Akt Inhibitor, Solid, 10 mg

Catalog No. 502583290
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3290 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3290 Supplier Abcam Supplier No. AB14375610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Highly validated small molecule Akt inhibitor.

  • Activity: Small molecule Akt inhibitor
  • Molecular Weight: 360.25 g/mol
  • Form: Solid
  • CAS Number: 956025-99-3
  • Citation Count: 11
  • Storage: Store at +4°C
  • Shipping: Ambient - Can Ship with Ice

Specifications

Form Solid
Shipping Condition Ambient
Target Molecule Akt
Content And Storage Store at 4°C.
Format Vial
Buffer 100% 3-[(3-bromophenyl)methyl]-1-tert-butylpyrazolo[3,4-d]pyrimidin-4-amine
Quantity 10 mg
For Use With (Application) Akt inhibitor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.