Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-1-(triisopropylsilyl)pyrrole 95.0+%, TCI America™
SDP

Catalog No. B18991G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18991G 1 g
B18995G 5 g
B189925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B18991G Supplier TCI America Supplier No. B18991G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 87630-36-2
Molecular Formula C13H24BrNSi
Molecular Weight (g/mol) 302.331
MDL Number MFCD01321136
InChI Key CNAUTMPRKBSDLH-UHFFFAOYSA-N
PubChem CID 2763302
IUPAC Name (3-bromopyrrol-1-yl)-tri(propan-2-yl)silane
SMILES CC(C)[Si](C(C)C)(C(C)C)N1C=CC(=C1)Br

Specifications

Color Yellow
Quantity 1 g
Formula Weight 302.33
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-1-(triisopropylsilyl)pyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.