Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Bromo-2,2':5',2″-terthiophene 97.0+%, TCI America™
SDP

Catalog No. B40501G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40501G 1 g
B40505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40501G Supplier TCI America Supplier No. B40501G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 105125-00-6
Molecular Formula C12H7BrS3
Molecular Weight (g/mol) 327.272
InChI Key FGBHDLKMGUOJBA-UHFFFAOYSA-N
Synonym 3-Bromo-2,5-di(2-thienyl)thiophene
PubChem CID 184442
IUPAC Name 3-bromo-2,5-dithiophen-2-ylthiophene
SMILES C1=CSC(=C1)C2=CC(=C(S2)C3=CC=CS3)Br

Specifications

Melting Point 40°C
Quantity 1 g
Formula Weight 327.27
Percent Purity ≥97.0% (GC)
Chemical Name or Material 3′-Bromo-2,2′:5′,2′′-terthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.