Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2,5-dichloropyridine 98.0+%, TCI America™
SDP

Catalog No. B48321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48321G 1 g
B48325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48321G Supplier TCI America Supplier No. B48321G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 138006-41-4
Molecular Formula C5H2BrCl2N
Molecular Weight (g/mol) 226.88
MDL Number MFCD06659484
InChI Key CDBHWRZEXFQNBI-UHFFFAOYSA-N
Synonym 2,5-dichloro-3-bromo-pyridine, 2-chloro-3-bromo-5-chloropyridine, 3-bromo-2,5-dichloro-pyridine, pyridine, 3-bromo-2,5-dichloro, 2,5-dichloro-3-bromopyridine, 2,5 dichloro-3-bromo-pyridine, pyridine,3-bromo-2,5-dichloro, pubchem1173, acmc-209cf8, 3-bromo-2, 5-dichloropyridine
PubChem CID 12901596
IUPAC Name 3-bromo-2,5-dichloropyridine
SMILES ClC1=CC(Br)=C(Cl)N=C1

Specifications

Melting Point 39°C
Color Yellow
Quantity 1 g
Formula Weight 226.88
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2,5-dichloropyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.