Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2,6-dimethoxytoluene 97.0+%, TCI America™
SDP

Catalog No. B47451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47451G 1 g
B4745200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47451G Supplier TCI America Supplier No. B47451G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22794-95-2
Molecular Formula C9H11BrO2
Molecular Weight (g/mol) 231.089
InChI Key WXHZGDILTIFQDK-UHFFFAOYSA-N
Synonym 1-Bromo-2,4-dimethoxy-3-methylbenzene
PubChem CID 10889743
IUPAC Name 1-bromo-2,4-dimethoxy-3-methylbenzene
SMILES CC1=C(C=CC(=C1OC)Br)OC

Specifications

Boiling Point 96°C
Quantity 1 g
Formula Weight 231.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-2,6-dimethoxytoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.