Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-chloro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Catalog No. AAH6443506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6443506 5 g
AAH6443503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6443506 Supplier Thermo Scientific Chemicals Supplier No. H6443506

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71701-92-3
Molecular Formula C6H2BrClF3N
Molecular Weight (g/mol) 260.438
MDL Number MFCD09878432
InChI Key GVQLMGPWTHAUPV-UHFFFAOYSA-N
Synonym 3-bromo-2-chloro-5-trifluoromethyl pyridine, 2-chloro-3-bromo-5-trifluoromethylpyridine, abbypharma ap-13-5058, pyridine, 3-bromo-2-chloro-5-trifluoromethyl, pubchem15158, acmc-1bcir, ksc496i9b, 2-morpholin-4-ylethanesulfonicacid,hydrate, 2-chloro-3-bromo-5-trifluoromethyl pyridine
PubChem CID 13536061
IUPAC Name 3-bromo-2-chloro-5-(trifluoromethyl)pyridine
SMILES C1=C(C=NC(=C1Br)Cl)C(F)(F)F

Specifications

Melting Point 28°C to 32°C
Color Brown to Yellow
Flash Point 98°C (208°F)
Quantity 5 g
UN Number UN2811
Formula Weight 260.44
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Bromo-2-chloro-5-(trifluoromethyl)pyridine
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.