Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-fluorobenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. B536325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B536325G 25 g
B53635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B536325G Supplier TCI America Supplier No. B536325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 149947-15-9
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD09027089
InChI Key OUAZPCKRSSEQKB-UHFFFAOYSA-N
Synonym 3-bromo-2-fluorobenzenecarbaldehyde, 3-bromo-2-fluorobenzadehyde, 2-fluoro-3-bromobenzaldehyde, benzaldehyde,3-bromo-2-fluoro, benzaldehyde, 3-bromo-2-fluoro, 3-bromofluorobenzaldehyde, intermediates-zcf02145, acmc-209d3i, 3-bromo-2-fluoro-benzaldehyde, 3-bromanyl-2-fluoranyl-benzaldehyde
PubChem CID 22019361
IUPAC Name 3-bromo-2-fluorobenzaldehyde
SMILES C1=CC(=C(C(=C1)Br)F)C=O

Specifications

Melting Point 43°C
Color White-Yellow
Quantity 25 g
Formula Weight 203.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2-fluorobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.