Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-fluorobenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. B51431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51431G 1 g
B51435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51431G Supplier TCI America Supplier No. B51431G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 261723-32-4
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.026
MDL Number MFCD09842441
InChI Key LIZLYZVAYZQVPG-UHFFFAOYSA-N
Synonym 3-bromo-2-fluorophenyl methanol, 1-bromo-2-fluoro-3-hydroxymethyl benzene, 3-bromo-2-fluorobenzyl alcohol, bromofluorophenylmethanol, benzenemethanol, 3-bromo-2-fluoro, benzenemethanol,3-bromo-2-fluoro, 3-bromo-2-fluoro-phenyl methanol, pubchem19843, acmc-209goz, ablock ab-12-8385
PubChem CID 26985646
IUPAC Name (3-bromo-2-fluorophenyl)methanol
SMILES C1=CC(=C(C(=C1)Br)F)CO

Specifications

Melting Point 38°C
Color White
Boiling Point 108°C
Quantity 1 g
Formula Weight 205.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2-fluorobenzyl Alcohol
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.