Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-furoyl chloride, 97%, Thermo Scientific™

Catalog No. CC67202DA
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC67202DA 1 g Amber Glass Bottle
CC67202DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC67202DA Supplier Thermo Scientific Chemicals Supplier No. CC67202DA
Add to Cart

Chemical Identifiers

CAS 58777-57-4
Molecular Formula C5H2BrClO2
Molecular Weight (g/mol) 209.423
MDL Number MFCD11109330
InChI Key WTZDPJVRZQGTET-UHFFFAOYSA-N
Synonym 3-bromo-2-furoyl chloride, 3-bromo-2-furancarbonyl chloride, 3-bromo-furan-2-carbonyl chloride, 2-furancarbonyl chloride, 3-bromo-9ci
PubChem CID 21864576
IUPAC Name 3-bromofuran-2-carbonyl chloride
SMILES C1=COC(=C1Br)C(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 209.43
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-furoyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.