Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-hydroxypyridine 97.0+%, TCI America™
SDP

Catalog No. B233025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B233025G 25 g
B23305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B233025G Supplier TCI America Supplier No. B233025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13466-43-8
Molecular Formula C5H4BrNO
Molecular Weight (g/mol) 174.00
MDL Number MFCD03411569
InChI Key YDUGVOUXNSWQSW-UHFFFAOYSA-N
Synonym 3-bromo-2-hydroxypyridine, 3-bromopyridin-2-ol, 3-bromopyridin-2 1h-one, 3-bromo-2-pyridinol, 3-bromo-2 1h-pyridinone, 2 1h-pyridinone, 3-bromo, 2-hydroxy-3-bromopyridine, 3-bromo-2 1h-pyridone, bromopyridone, 3-bromopyridone
PubChem CID 818549
IUPAC Name 3-bromo-1,2-dihydropyridin-2-one
SMILES BrC1=CC=CNC1=O

Specifications

Melting Point 182°C
Quantity 25 g
Formula Weight 174.00
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2-hydroxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.