Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-methoxypyridine 98.0+%, TCI America™
SDP

Catalog No. B37371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37371G 1 g
B37375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37371G Supplier TCI America Supplier No. B37371G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13472-59-8
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD03095349
InChI Key PORGLLGXCAQORO-UHFFFAOYSA-N
Synonym 2-methoxy-3-bromopyridine, 3-bromo-2-methoxy-pyridine, 3-brown-2-methoxypyridine, pyridine, 3-bromo-2-methoxy, tspp decahydrate, pubchem1180, 2-methoxybromopyridine, acmc-209bwq, ksc494g7f, 3-bromanyl-2-methoxy-pyridine
PubChem CID 12533390
IUPAC Name 3-bromo-2-methoxypyridine
SMILES COC1=C(Br)C=CC=N1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 188.02
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Bromo-2-methoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.