Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(-)-3-Bromo-2-Methyl-1-Propanol

Catalog No. 3250665G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-506-65G 5 g
32-506-61G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 32-506-65G Supplier Sigma Aldrich Supplier No. 3250665G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(-)-3-Bromo-2-methyl-1-propanol is used as a starting material in the total syntheses of epothilone C and bistramide A. It can also be used as a structural modifier in the homochiral porous molecular networks.

Specifications

CAS 93381-28-3
Density 1.461 g/mL at 20°C
Boiling Point 73°C to 74°C (9 mmHg, lit.)
Molecular Formula C4H9BrO
Refractive Index n20/D 1.484
Linear Formula BrCH2CH(CH3)CH2OH
MDL Number MFCD00013302
Quantity 5 g
Molecular Weight (g/mol) 153.02 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.