Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-fluorobenzoic Acid 96.0+%, TCI America™
SDP

Catalog No. B21331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21331G 1 g
B21335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21331G Supplier TCI America Supplier No. B21331G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1007-16-5
Molecular Formula C7H3BrFO2
Molecular Weight (g/mol) 218.00
MDL Number MFCD00042463
InChI Key ONELILMJNOWXSA-UHFFFAOYSA-M
Synonym 3-bromo-4-fluoro-benzoic acid, 4-fluoro-3-bromo benzoic acid, 3-bromo-4-fluorobenzoicacid, rarechem al bo 0604, buttpark 20\01-58, benzoic acid, 3-bromo-4-fluoro, 3-bromo-4-fluorbenzoic acid, pubchem1320, 3-bromo-4-fiuorobenzoic acid, 5-bromo-4-fluorobenzoic acid
PubChem CID 70509
IUPAC Name 3-bromo-4-fluorobenzoate
SMILES [O-]C(=O)C1=CC=C(F)C(Br)=C1

Specifications

Melting Point 139°C
Color White
Quantity 1 g
Formula Weight 219.01
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-fluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.