Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-fluorobenzotrifluoride 96.0+%, TCI America™
SDP

Catalog No. B14211G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14211G 1 g
B14215G 5 g
B142125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B14211G Supplier TCI America Supplier No. B14211G

Chemical Identifiers

CAS 68322-84-9
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 243.00
MDL Number MFCD00040938
InChI Key RZJOIMPUMMQKFR-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorobenzotrifluoride, 2-bromo-1-fluoro-4-trifluoromethyl benzene, 2-fluoro-5-trifluoromethyl bromobenzene, 3-bromo-alpha,alpha,alpha,4-tetrafluorotoluene, benzene, 2-bromo-1-fluoro-4-trifluoromethyl, 1-bromo-2-fluoro-5-trifluoromethyl benzene, 3-bromo-alpha,alpha,alpha-4-tetrafluorotoluene, pubchem1639, 3-bromo-4-fluoro trifluoromethyl benzene
PubChem CID 144295
IUPAC Name 2-bromo-1-fluoro-4-(trifluoromethyl)benzene
SMILES FC1=CC=C(C=C1Br)C(F)(F)F

Specifications

Color Yellow
Boiling Point 60°C
Quantity 1 g
Formula Weight 243.00
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-4-fluorobenzotrifluoride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.