Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-hydroxybenzonitrile 95.0+%, TCI America™
SDP

Catalog No. B203725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B203725G 25 g
B20375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B203725G Supplier TCI America Supplier No. B203725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2315-86-8
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.02
MDL Number MFCD00143096
InChI Key HLHNOIAOWQFNGW-UHFFFAOYSA-N
Synonym 2-bromo-4-cyanophenol, 3-bromo-4-hydroxy-benzonitrile, benzonitrile, 3-bromo-4-hydroxy, 3-bromo-4-hydroxybenzenecarbonitrile, timtec-bb sbb005818, labotest-bb lt01143437, pubchem3780, intermediates-zcf02624, acmc-209g1r
PubChem CID 75341
IUPAC Name 3-bromo-4-hydroxybenzonitrile
SMILES OC1=CC=C(C=C1Br)C#N

Specifications

Melting Point 156°C
Quantity 25 g
Formula Weight 198.02
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-hydroxybenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.