Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-methoxybenzyl alcohol, 98%, Thermo Scientific™

Catalog No. AAH32395MD
Change view
Click to view available options
Quantity:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32395MD 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH32395MD Supplier Thermo Scientific Chemicals Supplier No. H32395MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 38493-59-3
Molecular Formula C8H9BrO2
Quantity 250mg
MDL Number MFCD01861398
Synonym 3-bromo-4-methoxyphenyl methanol, 3-bromo-4-methoxybenzyl alcohol, benzenemethanol, 3-bromo-4-methoxy, ksc580a6r, 3-bromo4-methoxybenzyl alcohol, 3-bromo-4-methoxy-phenyl-methanol
InChI Key QOUYJSKYVZRGCV-UHFFFAOYSA-N
SMILES COC1=C(Br)C=C(CO)C=C1
IUPAC Name (3-bromo-4-methoxyphenyl)methanol
Molecular Weight (g/mol) 217.06
PubChem CID 13551346
Formula Weight 217.06
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-methoxybenzyl alcohol
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.