Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Bromo-4'-methylacetophenone 98.0+%, TCI America™
SDP

Catalog No. B47581G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47581G 1 g
B47585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47581G Supplier TCI America Supplier No. B47581G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 40180-80-1
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
MDL Number MFCD02683854
InChI Key PDJLFESXPVLVMR-UHFFFAOYSA-N
PubChem CID 3309401
IUPAC Name 1-(3-bromo-4-methylphenyl)ethanone
SMILES CC1=C(C=C(C=C1)C(=O)C)Br

Specifications

Melting Point 44°C
Color White-Yellow
Boiling Point 118°C
Quantity 1 g
Formula Weight 213.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Bromo-4′-methylacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.