Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-methylbenzaldehyde 96.0+%, TCI America™
SDP

Catalog No. B50731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50731G 1 g
B50735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50731G Supplier TCI America Supplier No. B50731G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 36276-24-1
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.047
MDL Number MFCD04971197
InChI Key WTXXUAHMTVAQHW-UHFFFAOYSA-N
PubChem CID 816765
IUPAC Name 3-bromo-4-methylbenzaldehyde
SMILES CC1=C(C=C(C=C1)C=O)Br

Specifications

Melting Point 48°C
Color White
Quantity 1 g
Formula Weight 199.05
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-methylbenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.