Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Bromo-5'-chloro-2'-hydroxyacetophenone 97.0+%, TCI America™
SDP

Catalog No. B223625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B223625G 25 g
B22365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B223625G Supplier TCI America Supplier No. B223625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59443-15-1
Molecular Formula C8H6BrClO2
Molecular Weight (g/mol) 249.49
MDL Number MFCD00857068
InChI Key FFAVKFQPEOGJOA-UHFFFAOYSA-N
Synonym 2-Acetyl-6-bromo-4-chlorophenol
PubChem CID 2735546
IUPAC Name 1-(3-bromo-5-chloro-2-hydroxyphenyl)ethan-1-one
SMILES CC(=O)C1=CC(Cl)=CC(Br)=C1O

Specifications

Melting Point 100°C
Color Yellow
Quantity 25 g
Formula Weight 249.49
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Bromo-5′-chloro-2′-hydroxyacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.