Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-chlorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. B41911G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41911G 1 g
B41915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41911G Supplier TCI America Supplier No. B41911G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42860-02-6
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.46
MDL Number MFCD06797227
InChI Key PBRRUJWXRUGGAW-UHFFFAOYSA-N
PubChem CID 33128
IUPAC Name 3-bromo-5-chlorobenzoic acid
SMILES OC(=O)C1=CC(Cl)=CC(Br)=C1

Specifications

Melting Point 197°C
Color White-Yellow
Quantity 1 g
Formula Weight 235.46
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-chlorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.