Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-chlorosalicylaldehyde 96.0+%, TCI America™
SDP

Catalog No. B194825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B194825G 25 g
B19485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B194825G Supplier TCI America Supplier No. B194825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19652-32-5
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.461
MDL Number MFCD00051690
InChI Key KNOYZLVIXXBBIB-UHFFFAOYSA-N
Synonym 3-bromo-5-chlorosalicylaldehyde, 3-bromo-5-chlorosalicylic aldehyde, benzaldehyde, 3-bromo-5-chloro-2-hydroxy, 3-bromo-5-chloro salicyclic aldehyde, 3-bromo-5-chloro-2-hydroxy-benzaldehyde, 3-bromo-5-chloro-o-hydroxybenzaldehyde, pubchem3591, acmc-1bs9s, akos bb-6948, knoyzlvixxbbib-uhfffaoysa
PubChem CID 519676
IUPAC Name 3-bromo-5-chloro-2-hydroxybenzaldehyde
SMILES C1=C(C=C(C(=C1Br)O)C=O)Cl

Specifications

Melting Point 88°C
Quantity 25 g
Formula Weight 235.46
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-chlorosalicylaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.