Learn More
3-Bromo-5-iodobenzoic acid, 97%
Description
3-Bromo-5-iodobenzoic acid may be used in the preparation of the following:
- Phenyl(3-bromo-5-iodo)benzoate.
- As starting reagent for the large-scale synthesis of the thromboxane receptor antagonist 3-{3-[2-(4-chlorobenzenesulfonamido)ethyl]-5-(4-fluorobenzyl)phenyl}propionic acid, via regioselective Heck cross-coupling reaction.
- Methyl 3-bromo-5-iodobenzoate.
- 3-Bromo-5-(triisopropylsilylethynyl)benzoic acid, via Sonogashira coupling reaction.
- Trifluoroacetophenone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Bromo-5-iodobenzoic acid may be used in the preparation of the following: • Phenyl(3-bromo-5-iodo)benzoate. • As starting reagent for the large-scale synthesis of the thromboxane receptor antagonist 3-{3-[2-(4-chlorobenzenesulfonamido)ethyl]-5-(4-fluorobenzyl)phenyl}propionic acid, via regioselective Heck cross-coupling reaction. • Methyl 3-bromo-5-iodobenzoate. • 3-Bromo-5-(triisopropylsilylethynyl)benzoic acid, via Sonogashira coupling reaction. • Trifluoroacetophenone.
Solubility
Soluble in methanol. Sparingly Soluble (0.20 g/L) (25°C), Calc.
Notes
Light Sensitive. Protect from light. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 218°C to 221°C |
| Quantity | 25 g |
| Beilstein | 3240837 |
| Sensitivity | Light sensitive |
| Solubility Information | Soluble in methanol. Sparingly Soluble (0.20g/L) (25°C),Calc. |
| Formula Weight | 326.92 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-Bromo-5-iodobenzoic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.