Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. B37241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37241G 1 g
B37245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37241G Supplier TCI America Supplier No. B37241G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3430-16-8
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD01646141
InChI Key ADCLTLQMVAEBLB-UHFFFAOYSA-N
Synonym 5-bromo-3-picoline, 3-bromo-5-picoline, 3-bromo-5-methyl-pyridine, 3-methyl-5-bromopyridine, pyridine, 3-bromo-5-methyl, 5-bromo-3-methylpyridine, 3-brom-5-methylpyridin, zlchem 381, 5-bromo-3-picolin, pubchem4036
PubChem CID 817713
IUPAC Name 3-bromo-5-methylpyridine
SMILES CC1=CC(Br)=CN=C1

Specifications

Color Yellow
Boiling Point 110°C
Quantity 1 g
Formula Weight 172.03
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Bromo-5-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.