Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-nitrobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. B33701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33701G 1 g
B33705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33701G Supplier TCI America Supplier No. B33701G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6307-83-1
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.016
MDL Number MFCD00100098
InChI Key AXRKIZCFYZBBPX-UHFFFAOYSA-N
Synonym 3-bromo-5-nitrobenzoic acid, 5-bromo-3-nitrobenzoic acid, benzoic acid, 3-bromo-5-nitro, 3-bromo-5-nitro-benzoic acid, 3-bromo-5-nitrobenzoicacid, pubchem4734, acmc-209ncw, ksc357s4t, 3-bromo-5-nitro benzoic acid
PubChem CID 239336
SMILES C1=C(C=C(C=C1[N+](=O)[O-])Br)C(=O)O

Specifications

Melting Point 162°C
Quantity 1 g
Formula Weight 246.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-5-nitrobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.