Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-6-fluoro-2-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. B513525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B513525G 25 g
B51355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B513525G Supplier TCI America Supplier No. B513525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 375368-83-5
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.02
MDL Number MFCD03095092
InChI Key GUYGQQWIKZLHTP-UHFFFAOYSA-N
Synonym 3-Bromo-6-fluoro-2-picoline
PubChem CID 17750149
IUPAC Name 3-bromo-6-fluoro-2-methylpyridine
SMILES CC1=C(Br)C=CC(F)=N1

Specifications

Color Colorless
Quantity 25 g
Formula Weight 190.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-6-fluoro-2-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.