Learn More
Apexbio Technology LLC 3-Bromo-7-nitroindazole 74209-34-0 50mg
SDP
Supplier:  Apexbio Technology LLC B642650
Encompass_Preferred
3-Bromo-7-nitroindazole is an indazole derivative structurally defined by a bromine and a nitro group substituted on the indazole scaffold It acts predominantly as a selective inhibitor of neuronal nitric oxide synthase (nNOS) thereby modulating the synthesis of nitric oxide in neural tissues Through nNOS inhibition 3-Bromo-7-nitroindazole is employed in biomedical research to investigate the physiological and pathological roles of nitric oxide particularly in studies related to neurodegeneration ischemia and inflammatory neural processes
Catalog No. 50-000-91777
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.