Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-9-n-octyl-9H-carbazole 98.0+%, TCI America™
SDP

Catalog No. B56101G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56101G 1 g
B56105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56101G Supplier TCI America Supplier No. B56101G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 628337-00-8
Molecular Formula C20H24BrN
Molecular Weight (g/mol) 358.323
InChI Key OQZRYECWBHQQAB-UHFFFAOYSA-N
PubChem CID 69630630
IUPAC Name 3-bromo-9-octylcarbazole
SMILES CCCCCCCCN1C2=C(C=C(C=C2)Br)C3=CC=CC=C31

Specifications

Melting Point 50°C
Boiling Point 135°C
Quantity 1 g
Formula Weight 358.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-9-n-octyl-9H-carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.