Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-p-anisaldehyde 98.0+%, TCI America™
SDP

Catalog No. B179025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B179025G 25 g
B17905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B179025G Supplier TCI America Supplier No. B179025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34841-06-0
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00016599
InChI Key QMPNFQLVIGPNEI-UHFFFAOYSA-N
Synonym 3-bromo-p-anisaldehyde, benzaldehyde, 3-bromo-4-methoxy, 3-bromo-4-methoxy-benzaldehyde, 3-bromoanisaldehyde, 5-bromoanisaldehyde, 3-bromo-anisaldehyde, 5-bromo anisaldehyde, pubchem2643, acmc-209iat, 2-bromo-4-formylanisole
PubChem CID 98662
IUPAC Name 3-bromo-4-methoxybenzaldehyde
SMILES COC1=CC=C(C=O)C=C1Br

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 162°C
Quantity 25 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-p-anisaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.