Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromobenzo[b]thiophene 96.0+%, TCI America™
SDP

Catalog No. B558925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B558925G 25 g
B55895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B558925G Supplier TCI America Supplier No. B558925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7342-82-7
Molecular Formula C8H5BrS
Molecular Weight (g/mol) 213.092
MDL Number MFCD00023009
InChI Key SRWDQSRTOOMPMO-UHFFFAOYSA-N
Synonym 3-bromobenzo b thiophene, 3-bromothianaphthene, 3-bromobenzothiophene, benzo b thiophene, 3-bromo, 3-bromo-benzo b thiophene, 3-bromo benzothiophene, benzo b thiophene, 3-bromo-,, zlchem 804
PubChem CID 123250
IUPAC Name 3-bromo-1-benzothiophene
SMILES C1=CC=C2C(=C1)C(=CS2)Br

Specifications

Boiling Point 269°C
Quantity 25 g
Formula Weight 213.09
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromobenzo[b]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.