Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. B0551250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0551250G 250 g
B055125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0551250G Supplier TCI America Supplier No. B0551250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 585-76-2
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.019
MDL Number MFCD00002487
InChI Key VOIZNVUXCQLQHS-UHFFFAOYSA-N
Synonym m-bromobenzoic acid, 3-brombenzoic acid, benzoic acid, 3-bromo, 3-bromo-benzoic acid, benzoic acid, m-bromo, unii-7dct3a73lx, 7dct3a73lx, 3-bromo benzene acid, 3-bromobenzoicacid, 4-fluorobenzoicacid
PubChem CID 11456
IUPAC Name 3-bromobenzoic acid
SMILES C1=CC(=CC(=C1)Br)C(=O)O

Specifications

Melting Point 158°C
Color White-Yellow
Quantity 250 g
Formula Weight 201.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromobenzoic Acid
EINECSNumber (3)-0062
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.