Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromobenzotrifluoride 99.0+%, TCI America™
SDP

Catalog No. B0664250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0664250G 250 g
B066425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0664250G Supplier TCI America Supplier No. B0664250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 401-78-5
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.008
MDL Number MFCD00000380
InChI Key NNMBNYHMJRJUBC-UHFFFAOYSA-N
Synonym 3-bromobenzotrifluoride, 1-bromo-3-trifluoromethyl benzene, m-bromobenzotrifluoride, 3-trifluoromethyl bromobenzene, benzene, 1-bromo-3-trifluoromethyl, 3-brombenzotrifluorid, 3-bromotrifluoromethylbenzene, 3-bromobenzyltrifluoride, 3-trifluoromethyl phenyl bromide, m-trifluoromethyl bromobenzene
PubChem CID 9817
IUPAC Name 1-bromo-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)Br)C(F)(F)F

Specifications

Boiling Point 155°C
Quantity 250 g
UN Number 1993
Formula Weight 225.01
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromobenzotrifluoride
EINECSNumber (3)-0068&(9)-1242
RTECSNumber XS7970000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.