Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromobenzylamine, 95%

Catalog No. AAB2496314
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2496314 25 g
AAB2496306 5 g
AAB2496322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2496314 Supplier Thermo Scientific Chemicals Supplier No. B2496314
Only null left
Add to Cart

CAS: 10269-01-9 | C7H8BrN | 186.052 g/mol

3-Bromobenzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromobenzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10269-01-9
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD01026119
InChI Key SUYJXERPRICYRX-UHFFFAOYSA-N
Synonym 3-bromobenzylamine, 3-bromophenyl methanamine, benzenemethanamine, 3-bromo, 3-bromophenyl methylamine, m-bromobenzylamine, 1-3-bromophenyl methanamine, 3-bromo-benzylamine, 3-bromobenzenemethanamine, 3-bromobenzyl amine, pubchem21109
PubChem CID 457587
IUPAC Name (3-bromophenyl)methanamine
SMILES C1=CC(=CC(=C1)Br)CN

Specifications

Odor Amine-like
Refractive Index 1.5885
Quantity 25 g
UN Number UN2735
Sensitivity Air sensitive
Solubility Information Slightly soluble in water.
Formula Weight 186.06
Percent Purity 97%
Chemical Name or Material 3-Bromobenzylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 679-024-4
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.