Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromobiphenyl, TCI America™
SDP

Catalog No. B200125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B200125G 25 g
B20011G 1 g
B20015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B200125G Supplier TCI America Supplier No. B200125G

Chemical Identifiers

CAS 2113-57-7
Molecular Formula C12H9Br
Molecular Weight (g/mol) 233.11
MDL Number MFCD00000082
InChI Key USYQKCQEVBFJRP-UHFFFAOYSA-N
Synonym 3-bromobiphenyl, 3-bromo-1,1'-biphenyl, 1,1'-biphenyl, 3-bromo, 3-bromo-biphenyl, m-bromobiphenyl, biphenyl, 3-bromo, 3-bromodiphenyl, unii-o7i53k56s8, ccris 5889, 3-brombiphenyl
PubChem CID 16449
IUPAC Name 3-bromo-1,1'-biphenyl
SMILES BrC1=CC=CC(=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 301°C
Quantity 25 g
Formula Weight 233.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromobiphenyl
EINECSNumber (4)-0018
RTECSNumber DV1750000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.