Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromocarbazole 97.0+%, TCI America™
SDP

Catalog No. B345825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B345825G 25 g
B34585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B345825G Supplier TCI America Supplier No. B345825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1592-95-6
Molecular Formula C12H8BrN
Molecular Weight (g/mol) 246.107
MDL Number MFCD00222621
InChI Key LTBWKAYPXIIVPC-UHFFFAOYSA-N
Synonym 3-bromocarbazole, 9h-carbazole, 3-bromo, 3-brominecarbazole, 3bromo-9h-carbazole, pubchem9950, maybridge1_004311, acmc-1bpt1, 3-bromocarbazole gc
PubChem CID 252446
IUPAC Name 3-bromo-9H-carbazole
SMILES C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)Br

Specifications

Melting Point 198°C
Color White-Yellow
Boiling Point 160°C
Quantity 25 g
Formula Weight 246.11
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromocarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.