Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromocyclohexene, 95%

Catalog No. p-7080724
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1037006 5 g
AAA1037014 25 g
AAA1037022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1037006 Supplier Thermo Scientific Chemicals Supplier No. A1037006
Only null left
Add to Cart

CAS: 1521-51-3 | C6H9Br | 161.042 g/mol

In peptide synthesis, can be used to protect the hydroxyl group of serine and threonine as the cyclohexyl (Chx) ether, by reaction with the sodium alkoxide (generated with NaH), followed by hydrogenation. The group is stable to TFA and 20% piperidine in DMF, but can be cleaved with TfOH in TFA, in the presence of thioanisole as a cation scavenger. It is also used in the synthesis of N-tert-butoxycarbonyl-O-cyclohexyl-L-tyrosine.1 It was also used in the synthesis of enantiopure cyclohexitols such as muco-quercitol, D-chiro-inocitol and allo-inocitol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In peptide synthesis, can be used to protect the hydroxyl group of serine and threonine as the cyclohexyl (Chx) ether, by reaction with the sodium alkoxide (generated with NaH), followed by hydrogenation. The group is stable to TFA and 20% piperidine in DMF, but can be cleaved with TfOH in TFA, in the presence of thioanisole as a cation scavenger. It is also used in the synthesis of N-tert-butoxycarbonyl-O-cyclohexyl-L-tyrosine.1 It was also used in the synthesis of enantiopure cyclohexitols such as muco-quercitol, D-chiro-inocitol and allo-inocitol.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1521-51-3
Molecular Formula C6H9Br
Molecular Weight (g/mol) 161.042
MDL Number MFCD00013775
InChI Key AJKDUJRRWLQXHM-UHFFFAOYSA-N
Synonym 3-bromocyclohex-1-ene, cyclohexene, 3-bromo, 1-bromo-2-cyclohexene, 3-bromo-1-cyclohexene, 1,2,3,4-tetrahydrobromobenzene, 3-bromo-cyclohexene, 2-cyclohexenyl bromide, 2-cyclohexen-1-yl bromide, 3-bromo cyclohexene, 3-bromanylcyclohexene
PubChem CID 137057
IUPAC Name 3-bromocyclohexene
SMILES C1CC=CC(C1)Br

Specifications

Density 1.4
Boiling Point 64°C to 65°C (15 mmHg)
Flash Point 54°C (129°F)
Refractive Index 1.531
Quantity 5 g
UN Number UN1993
Beilstein 635953
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 161.05
Percent Purity 95%
Chemical Name or Material 3-Bromocyclohexene, Stabilized with 300 to 1000ppm Propylene oxide
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.