Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 3-Bromocytisine
GREENER_CHOICESDP

Catalog No. 354950
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-495-0 50 mg
35-491-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-495-0 Supplier Tocris Bioscience™ Supplier No. 3549/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent agonist of α4β4, α4β2 and α7 nACh receptors

Potent agonist of α4β4, α4β2 and α7 nACh receptors (IC50 values are 0.28, 0.30 and 31.6 nM respectively). Displays different effects on high (HS) and low (LS) ACh sensitivity α4β2 nAChRs (EC50 values are 0.008 and 0.05 μM respectively).

Chemical Identifiers

CAS 207390-14-5
Molecular Formula C11H13BrN2O

Specifications

Quantity 50 mg
Formula Weight 269.14
Purity >98%
Molecular Weight (g/mol) 269.142
Chemical Name or Material 3-Bromocytisine
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.