Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromodibenzofuran 98.0+%, TCI America™
SDP

Catalog No. B54511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54511G 1 g
B5451200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B54511G Supplier TCI America Supplier No. B54511G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26608-06-0
Molecular Formula C12H7BrO
Molecular Weight (g/mol) 247.09
MDL Number MFCD09966042
InChI Key AZFABGHLDGJASW-UHFFFAOYSA-N
PubChem CID 296861
IUPAC Name 5-bromo-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
SMILES BrC1=CC2=C(C=C1)C1=C(O2)C=CC=C1

Specifications

Melting Point 123°C
Color White
Quantity 1 g
Formula Weight 247.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromodibenzofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.